Tisk…

Anti-CIITA Rabbit Polyclonal Antibody

Dodavatel: Abcam
ABCAAB7541-100EA 22310 CZK
ABCAAB7541-100
Anti-CIITA Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: CIITA (class II, major histocompatibility complex, transactivator)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR