Tisk…

Anti-APOA1 Goat Polyclonal Antibody

Dodavatel: Abcam
ABCAAB7614-250EA 23570 CZK
ABCAAB7614-250
Anti-APOA1 Goat Polyclonal Antibody
Protilátky
Type: Primary
Antigen: APOA1 (apolipoprotein A-I)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR