Tisk…

Anti-Cyanine Sheep Polyclonal Antibody (Biotin)

Dodavatel: Abcam
ABCAAB7625-100EA 21300 CZK
ABCAAB7625-100
Anti-Cyanine Sheep Polyclonal Antibody (Biotin)
Protilátky
Type: Primary
Antigen: Cyanine
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Sheep
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR