Tisk…

Anti-SERPINA1 Goat Polyclonal Antibody HRP (Horseradish Peroxidase)

ABCAAB7635-5EA 15650 CZK
ABCAAB7635-5
Anti-SERPINA1 Goat Polyclonal Antibody HRP (Horseradish Peroxidase)
Type: Primary
Antigen: SERPINA1 (serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 1)
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Goat
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR