Tisk…

Anti-Phosphoserine Mouse Monoclonal Antibody [clone: B106.1]

Dodavatel: Abcam
ABCAAB7851-500EA 22340 CZK
ABCAAB7851-500
Anti-Phosphoserine Mouse Monoclonal Antibody [clone: B106.1]
Protilátky
Type: Primary
Antigen: Phosphoserine
Clonality: monoclonal
Clone: B106.1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR