Tisk…

Anti-HLA DR + DP + DQ Mouse Monoclonal Antibody [clone: CR3/43]

Dodavatel: Abcam
ABCAAB7856-500EA 22830 CZK
ABCAAB7856-500
Anti-HLA DR + DP + DQ Mouse Monoclonal Antibody [clone: CR3/43]
Protilátky
Type: Primary
Antigen: HLA DR + DP + DQ
Clonality: monoclonal
Clone: CR3/43
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR