Tisk…

Anti-SRC Rabbit Polyclonal Antibody

ABCAAB7950EA 16680 CZK
ABCAAB7950
Anti-SRC Rabbit Polyclonal Antibody
Type: Primary
Antigen: SRC (v-src avian sarcoma (Schmidt-Ruppin A-2) viral oncogene homolog)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR