Tisk…

Anti-STAT5A Rabbit Polyclonal Antibody

ABCAAB7968-1EA 16280 CZK
ABCAAB7968-1
Anti-STAT5A Rabbit Polyclonal Antibody
Type: Primary
Antigen: STAT5A (signal transducer and activator of transcription 5A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR