Tisk…

Anti-CXCR6 Rabbit Polyclonal Antibody

Dodavatel: Abcam
ABCAAB8023-100EA 23040 CZK
ABCAAB8023-100
Anti-CXCR6 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: CXCR6 (chemokine (C-X-C motif) receptor 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR