Tisk…

Anti-BACE2 Rabbit Polyclonal Antibody

Dodavatel: Abcam
ABCAAB8024-100EA 21850 CZK
ABCAAB8024-100
Anti-BACE2 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: BACE2 (beta-site APP-cleaving enzyme 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR