Tisk…

Anti-BrdU Mouse Monoclonal Antibody [clone: MoBu-1]

Dodavatel: Abcam
ABCAAB8039-100EA 23320 CZK
ABCAAB8039-100
Anti-BrdU Mouse Monoclonal Antibody [clone: MoBu-1]
Protilátky
Type: Primary
Antigen: BrdU (Bromodeoxyuridine)
Clonality: monoclonal
Clone: MoBu-1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR