Tisk…
Anti-IP10 Mouse Monoclonal Antibody (Biotin) [clone: 6D4]
Dodavatel: Abcam
ABCAAB8099-50EA 25740 CZK
ABCAAB8099-50
Anti-IP10 Mouse Monoclonal Antibody (Biotin) [clone: 6D4]
Protilátky
Type: Primary
Antigen: IP10
Clonality: monoclonal
Clone: 6D4
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human