Tisk…

Anti-GPR15 Rabbit Polyclonal Antibody

Dodavatel: Abcam
ABCAAB8104-100EA 27030 CZK
ABCAAB8104-100
Anti-GPR15 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: GPR15 (G protein-coupled receptor 15)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR