Tisk…

Anti-pro Caspase 12 Rabbit Polyclonal Antibody

Dodavatel: Abcam
ABCAAB8118-100EA 27460 CZK
ABCAAB8118-100
Anti-pro Caspase 12 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: pro Caspase 12
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR