Tisk…

Anti-p60 CAF1 Mouse Monoclonal Antibody [clone: SS 53]

ABCAAB8133-100EA 16480 CZK
ABCAAB8133-100
Anti-p60 CAF1 Mouse Monoclonal Antibody [clone: SS 53]
Type: Primary
Antigen: p60 CAF1
Clonality: monoclonal
Clone: SS 53
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR