Tisk…

Anti-ACTA2 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 1A4]

Dodavatel: Abcam
ABCAAB8211-100EA 30950 CZK
ABCAAB8211-100
Anti-ACTA2 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 1A4]
Protilátky
Type: Primary
Antigen: ACTA2 (actin, alpha 2, smooth muscle, aorta)
Clonality: monoclonal
Clone: 1A4
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR