Tisk…

Anti-Adenovirus hexon protein Mouse Monoclonal Antibody [clone: 1E11]

Dodavatel: Abcam
ABCAAB8251-200EA 21140 CZK
ABCAAB8251-200
Anti-Adenovirus hexon protein Mouse Monoclonal Antibody [clone: 1E11]
Protilátky
Type: Primary
Antigen: Adenovirus hexon protein
Clonality: monoclonal
Clone: 100000000000
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR