Tisk…

Anti-Rubella Virus structural glycoprotein Mouse Monoclonal Antibody [clone: Ru6]

ABCAAB8267-200EA 16360 CZK
ABCAAB8267-200
Anti-Rubella Virus structural glycoprotein Mouse Monoclonal Antibody [clone: Ru6]
Type: Primary
Antigen: Rubella Virus structural glycoprotein
Clonality: monoclonal
Clone: Ru6
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR