Tisk…

Anti-Toxoplasma gondii Mouse Monoclonal Antibody [clone: TP3]

Dodavatel: Abcam
ABCAAB8313-250EA 21330 CZK
ABCAAB8313-250
Anti-Toxoplasma gondii Mouse Monoclonal Antibody [clone: TP3]
Protilátky
Type: Primary
Antigen: Toxoplasma gondii
Clonality: monoclonal
Clone: TP3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR