Tisk…

Anti-GABRB2 Rabbit Polyclonal Antibody

Dodavatel: Abcam
ABCAAB8340-100EA 21140 CZK
ABCAAB8340-100
Anti-GABRB2 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: GABRB2 (gamma-aminobutyric acid (GABA) A receptor, beta 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR