Tisk…

Anti-HIF1A Mouse Monoclonal Antibody [clone: ESEE122]

Dodavatel: Abcam
ABCAAB8366-100EA 23870 CZK
ABCAAB8366-100
Anti-HIF1A Mouse Monoclonal Antibody [clone: ESEE122]
Protilátky
Type: Primary
Antigen: HIF1A (hypoxia inducible factor 1, alpha subunit (basic helix-loop-helix transcription factor))
Clonality: monoclonal
Clone: ESEE122
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR