Tisk…

Anti-TNFRSF25 Rabbit Polyclonal Antibody

Dodavatel: Abcam
ABCAAB8412-100EA 23080 CZK
ABCAAB8412-100
Anti-TNFRSF25 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: TNFRSF25 (tumor necrosis factor receptor superfamily, member 25)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR