Tisk…

Anti-TNFRSF10B Rabbit Polyclonal Antibody

Dodavatel: Abcam
ABCAAB8416-100EA 23840 CZK
ABCAAB8416-100
Anti-TNFRSF10B Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: TNFRSF10B (tumor necrosis factor receptor superfamily, member 10b)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR