Tisk…

Anti-TNFRSF21 Rabbit Polyclonal Antibody

Dodavatel: Abcam
ABCAAB8417-100EA 22520 CZK
ABCAAB8417-100
Anti-TNFRSF21 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: TNFRSF21 (tumor necrosis factor receptor superfamily, member 21)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR