Tisk…

Anti-FLIP delta + gamma Rabbit Polyclonal Antibody

Dodavatel: Abcam
ABCAAB8423-100EA 20380 CZK
ABCAAB8423-100
Anti-FLIP delta + gamma Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: FLIP delta + gamma
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR