Tisk…

Anti-IgM Goat Polyclonal Antibody

Dodavatel: Abcam
ABCAAB8496-1EA 8460 CZK
ABCAAB8496-1
Anti-IgM Goat Polyclonal Antibody
Protilátky
Type: Secondary
Antigen: IgM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR