Tisk…

Anti-IgA/IgG/IgM Rabbit Polyclonal Antibody

Dodavatel: Abcam
ABCAAB8516-1EA 9040 CZK
ABCAAB8516-1
Anti-IgA/IgG/IgM Rabbit Polyclonal Antibody
Protilátky
Type: Secondary
Antigen: Mouse IgG+IgM+IgA
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR