Tisk…

Anti-Hepatitis B Virus Surface Antigen Mouse Monoclonal Antibody [clone: S14]

Dodavatel: Abcam
ABCAAB8636-100EA 22650 CZK
ABCAAB8636-100
Anti-Hepatitis B Virus Surface Antigen Mouse Monoclonal Antibody [clone: S14]
Protilátky
Type: Primary
Antigen: Hepatitis B Virus Surface Antigen
Clonality: monoclonal
Clone: S14
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR