Tisk…

Anti-Hepatitis B Virus Core Antigen Mouse Monoclonal Antibody [clone: C1]

Dodavatel: Abcam
ABCAAB8637-100EA 23600 CZK
ABCAAB8637-100
Anti-Hepatitis B Virus Core Antigen Mouse Monoclonal Antibody [clone: C1]
Protilátky
Type: Primary
Antigen: Hepatitis B Virus Core Antigen
Clonality: monoclonal
Clone: C1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR