Tisk…

Anti-C1R Sheep Polyclonal Antibody

ABCAAB8781-1EA 15650 CZK
ABCAAB8781-1
Anti-C1R Sheep Polyclonal Antibody
Type: Primary
Antigen: C1R (complement component 1, r subcomponent)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR