Tisk…

Anti-CTSG Sheep Polyclonal Antibody

ABCAAB8816-1EA 15650 CZK
ABCAAB8816-1
Anti-CTSG Sheep Polyclonal Antibody
Type: Primary
Antigen: CTSG (cathepsin G)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR