Tisk…

Anti-Factor XIII Sheep Polyclonal Antibody

ABCAAB8828-1EA 15370 CZK
ABCAAB8828-1
Anti-Factor XIII Sheep Polyclonal Antibody
Type: Primary
Antigen: Factor XIII
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR