Tisk…

Anti-SOD2 Sheep Polyclonal Antibody

ABCAAB8867-1EA 15370 CZK
ABCAAB8867-1
Anti-SOD2 Sheep Polyclonal Antibody
Type: Primary
Antigen: SOD2 (superoxide dismutase 2, mitochondrial)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR