Tisk…

Anti-p75 NGF Receptor Rabbit Polyclonal Antibody

ABCAAB8875-50EA 19770 CZK
ABCAAB8875-50
Anti-p75 NGF Receptor Rabbit Polyclonal Antibody
Type: Primary
Antigen: p75 NGF Receptor
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR