Tisk…

Anti-Dopamine Mouse Monoclonal Antibody [clone: HYDA001]

ABCAAB8892-50EA 16480 CZK
ABCAAB8892-50
Anti-Dopamine Mouse Monoclonal Antibody [clone: HYDA001]
Type: Primary
Antigen: Dopamine
Clonality: Monoclonal
Clone: HYDA001
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR