Tisk…

Anti-Notch 2 intracellular domain Rabbit Polyclonal Antibody

Dodavatel: Abcam
ABCAAB8927-200EA 22340 CZK
ABCAAB8927-200
Anti-Notch 2 intracellular domain Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: Notch 2 intracellular domain
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR