Tisk…

Anti-APOB Sheep Polyclonal Antibody HRP (Horseradish Peroxidase)

ABCAAB8948-1EA 15650 CZK
ABCAAB8948-1
Anti-APOB Sheep Polyclonal Antibody HRP (Horseradish Peroxidase)
Type: Primary
Antigen: APOB (apolipoprotein B)
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Sheep
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR