Tisk…

Anti-Cardiotin Mouse Monoclonal Antibody [clone: SR-2]

ABCAAB8963-100EA 15370 CZK
ABCAAB8963-100
Anti-Cardiotin Mouse Monoclonal Antibody [clone: SR-2]
Type: Primary
Antigen: Cardiotin
Clonality: monoclonal
Clone: SR-2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human, Pig
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR