Tisk…

Anti-VIM Mouse Monoclonal Antibody [clone: RV202]

Dodavatel: Abcam
ABCAAB8978-100EA 33090 CZK
ABCAAB8978-100
Anti-VIM Mouse Monoclonal Antibody [clone: RV202]
Protilátky
Type: Primary
Antigen: VIM (vimentin)
Clonality: monoclonal
Clone: RV202
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR