Tisk…

Anti-Lactate Dehydrogenase Isoenzyme V Sheep Polyclonal Antibody

ABCAAB9002-1EA 16880 CZK
ABCAAB9002-1
Anti-Lactate Dehydrogenase Isoenzyme V Sheep Polyclonal Antibody
Type: Primary
Antigen: Lactate Dehydrogenase Isoenzyme V
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR