Tisk…

Anti-Lambda Light chain Sheep Polyclonal Antibody (FITC (Fluorescein))

Dodavatel: Abcam
ABCAAB9006-1EA 21140 CZK
ABCAAB9006-1
Anti-Lambda Light chain Sheep Polyclonal Antibody (FITC (Fluorescein))
Protilátky
Type: Primary
Antigen: Lambda Light chain
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Sheep
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR