Tisk…

Anti-TF Sheep Polyclonal Antibody

Dodavatel: Abcam
ABCAAB9033-1EA 21640 CZK
ABCAAB9033-1
Anti-TF Sheep Polyclonal Antibody
Protilátky
Type: Primary
Antigen: TF (transferrin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR