Tisk…

Anti-NEFL Rabbit Polyclonal Antibody

Dodavatel: Abcam
ABCAAB9035-50EA 27760 CZK
ABCAAB9035-50
Anti-NEFL Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: NEFL (neurofilament, light polypeptide)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR