Tisk…

Anti-HTLV I gp46 Mouse Monoclonal Antibody [clone: 67/5.5.13.1]

Dodavatel: Abcam
ABCAAB9082-100EA 22860 CZK
ABCAAB9082-100
Anti-HTLV I gp46 Mouse Monoclonal Antibody [clone: 67/5.5.13.1]
Protilátky
Type: Primary
Antigen: HTLV I gp46
Clonality: monoclonal
Clone: 67/5.5.13.1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR