Tisk…

Anti-BRCA1 Rabbit Polyclonal Antibody

Dodavatel: Abcam
ABCAAB9141-50EA 23080 CZK
ABCAAB9141-50
Anti-BRCA1 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: BRCA1 (breast cancer 1, early onset)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR