Tisk…

Anti-Phosphoserine Rabbit Polyclonal Antibody

Dodavatel: Abcam
ABCAAB9332-100EA 23960 CZK
ABCAAB9332-100
Anti-Phosphoserine Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: Phosphoserine
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR