Tisk…
Anti-Phosphothreonine-X-Phosphotyrosine Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Dodavatel: Abcam
ABCAAB9342-100EA 20380 CZK
ABCAAB9342-100
Anti-Phosphothreonine-X-Phosphotyrosine Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Protilátky
Type: Primary
Antigen: Phosphothreonine-X-Phosphotyrosine
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Objednat