Tisk…

Anti-IL11 Rabbit Polyclonal Antibody

ABCAAB9552-100EA 19190 CZK
ABCAAB9552-100
Anti-IL11 Rabbit Polyclonal Antibody
Type: Primary
Antigen: IL11 (interleukin 11)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR