Tisk…

Anti-IL16 Rabbit Polyclonal Antibody

Dodavatel: Abcam
ABCAAB9563-100EA 21140 CZK
ABCAAB9563-100
Anti-IL16 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: IL16 (interleukin 16)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR