Tisk…

Anti-FLT3LG Rabbit Polyclonal Antibody

Dodavatel: Abcam
ABCAAB9688-100EA 21640 CZK
ABCAAB9688-100
Anti-FLT3LG Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: FLT3LG (fms-related tyrosine kinase 3 ligand)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR